ContactUsLinkComponent
Vi-CELL* Concentration Control, For daily check, Size: 1 Bottlex20ml
Vi-CELL Reagents and Consumables, Beckman Coulter
Catalog #: 76176-648
Supplier:  Beckman Coulter
Vi-CELL* Concentration Control, For daily check, Size: 1 Bottlex20ml
Vi-CELL Reagents and Consumables, Beckman Coulter
Catalog #: 76176-648
Supplier:  Beckman Coulter
Specifications
  • Description:
    Vi-CELL XR Concentration Control (Beads), 20 ml/bottle
  • Cat. no.:
    76176-648
Specifications
About this item
Consumables from the Vi-CELL manufacturer ensure optimum performance and results from your Vi-CELL.
  • Concentration Control to run daily to ensure proper instrument performance
  • Focus Control to perform auto-focus routine
  • Sample vials designed to properly fit in the Vi-CELL sample carousel
  • Reagent Paks designed to fit in the Vi-CELL XR with color-coded tubing for error-free installation
The Vi-CELL Concentration Control should be run daily to ensure proper instrument performance and results.
The Vi-CELL Focus Control should be used with the Focusing Wizard to automatically check and if necessary, adjust the focus; especially after the instrument has been physically moved to ensure optimum results.
The Vi-CELL sample vials, which hold up to 4 mL, should be used exclusively with the Vi-CELL, as they have been designed specifically for the carousel.
The Vi-CELL Quad Paks are the key to the Vi-CELL automated sample preparation for the Trypan Blue cell viability analysis . Each reagent pak is color coded to match the reagent lines to make replacing the pak easy.
Caution: Reagents and vials ship separately due to flammable materials. Please order vicell cups (76176-644) and reagents (76474-336) individually if you are in a GMP enviornment.
Customers Who Bought This Also Bought