Siliaprep* Spe Cartridge Tosic Acid (Scx) 2 G
Catalog #: CA75958-680
Supplier: SiliCycle
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Supplier: Ricca Chemical
$200.82 to $595.74
Sodium hydroxide 0.01 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$82.69 to $978.38
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$169.74 to $169.74
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$169.41 to $781.78
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61