SiliaPrep* SPE cartridgeAmine (WAX) nec 500mg
Catalog #: CA75958-558
Supplier: SiliCycle
Customers Who Bought This Also Bought
Iron, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
415,20 $
Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
103,36 $ to 1 240,08 $
Scalpel Handles; Feather®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
54,34 $
Ammonia solution 28.0 - 30.0% (NH₃ basis), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
917,17 $
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99%
Supplier: Thermo Scientific Chemicals
109,00 $