SiliaPrep* SPE cartridge Silica 1 g
Catalog #: CA75958-206
Supplier: SiliCycle
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Hydrogen peroxide 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Supplier: VWR International
$568.42
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Mix & Go, <i>E. coli</i> Transformation Kit & Buffer Set, Zymo Research
Supplier: Zymo Research
$204.14 to $204.14