SiliaPrepX* SPE cartridge WAX 30mg
Catalog #: CA75958-096
Supplier: SiliCycle
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Copan Regular Polyester Tip Swab with Plastic Applicator, Sterile, Hardy Diagnostics
Supplier: Hardy Diagnostics
$50.20