SiliaPrep* SPE cartridge Alumina-Basic 50 mg
Catalog #: CA75957-794
Supplier: SiliCycle
Customers Who Bought This Also Bought
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$741.97 to $47,885.70
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Supplier: Ricca Chemical
$200.82 to $595.74
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$260.48 to $788.04
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$78.90 to $874.91