SiliaChrom* Plus PFP HPLC Column 4.6 x 50 mm
Catalog #: CA75876-656
Supplier: SiliCycle
Customers Who Bought This Also Bought
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$72.06 to $624.50
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80
MinION™ DNA/RNA Sequencing Device, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
$3,314.84
Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$218.47 to $218.47
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$231.64
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$471.36 to $2,828.23