MiniVID USB 3.0 Super-Fast Camera 6.3MP
Catalog #: 470231-932
Supplier: LW Scientific
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$233.35 to $2,174.33
Anti-CD41/CD61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: PAC-1]
Supplier: Biolegend
$510.34
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$795.94 to $2,497.03