Order Entry
ContactUsLinkComponent
Quick-DNA* Fecal/Soil Microbe Miniprep Kit
Quick-DNA™ Fecal/Soil Microbe Kits, Zymo Research
Catalog #: 77001-814
Supplier:  Zymo Research
CAS Number:  
Quick-DNA* Fecal/Soil Microbe Miniprep Kit
Quick-DNA™ Fecal/Soil Microbe Kits, Zymo Research
Catalog #: 77001-814
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Stool
    Soil
  • Target molecule:
    DNA
  • Description:
    Quick-DNA™ Fecal/Soil Microbe Miniprep Kit
  • Size:
    50 preps
  • Description:
    Quick-DNA™ Fecal/Soil Microbe Miniprep Kit

Specifications

About this item

Rapid methods for the isolation of inhibitor-free, PCR quality DNA from fecal, soil, and microbial samples in minutes including tough-to-lyse bacteria, fungi, algae, and protozoa.

  • Ultra-high density BashingBeads™ are fracture resistant and chemically inert
  • Zymo-Spin™ Column and unique filtration technologies effectively removes PCR inhibitors from the DNA product

The Quick-DNA™ Fecal/Soil Microbe Kits are designed for the simple and rapid isolation of inhibitor-free, PCR-quality DNA from microbes in soil and feces. This kit can be used to isolate DNA from variety of soils (clay, sandy, silty, peaty, chalky, and loamy) and feces (humans, birds, rats, mice, cattle, etc.). Tough-to-lyse bacteria, fungi, protozoa, and algae are all rapidly and efficiently lysed by bead beating with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ Column or Plate technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. The procedures can be performed in minutes, and there is no need for organic denaturants or proteinases. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection, etc.

Customers Who Bought This Also Bought