Order Entry
ContactUsLinkComponent
Fungal/Bacterial Miniprep Kit 50 Preps
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Catalog #: 77001-808
Supplier:  Zymo Research
CAS Number:  
Fungal/Bacterial Miniprep Kit 50 Preps
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Catalog #: 77001-808
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Yeast/Fungal DNA
    Bacterial DNA
  • Sample type:
    Bacteria
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    Quick-DNA™ Fungal/Bacterial Miniprep Kit
  • Size:
    50 preps
  • Description:
    Quick-DNA™ Fungal/Bacterial Miniprep Kit

Specifications

About this item

Quick, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria.

  • Simple, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria in minutes
  • Ultra-high density BashingBeads™ are fracture resistant and chemically inert

The Quick-DNA™ Fungal/Bacterial Kits are designed for the simple and rapid isolation of DNA from tough-to-lyse fungi, including A. fumigatus, C. albicans, N. crassa, S. cerevisiae, S. pombe, as well as Gram-positive/negative bacteria, algae, and protozoa. The procedures are easy and can be completed in minutes. Fungal and/or bacterial samples are rapidly and efficiently lysed with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ technology is then used to isolate the DNA, which is ideal for downstream molecular-based applications including PCR, array, etc.

Customers Who Bought This Also Bought