Order Entry
ContactUsLinkComponent
ChIP DNA Clean and Concentrator 50 preps
ChIP DNA Clean and Concentrator™ Kits, Zymo Research
Catalog #: 77001-700
Supplier:  Zymo Research
CAS Number:  
ChIP DNA Clean and Concentrator 50 preps
ChIP DNA Clean and Concentrator™ Kits, Zymo Research
Catalog #: 77001-700
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Immunoprecipitates
  • Target molecule:
    DNA
  • Description:
    ChIP DNA Clean Concentrator Kit, Uncapped Column
  • Size:
    50 preps
  • Description:
    ChIP DNA Clean Concentrator Kit
  • Cat. no.:
    77001-700

Specifications

About this item

The Chromatin Immunoprecipitation (ChIP) DNA Clean & Concentrator provides a hassle-free method for the rapid purification and concentration of high-quality DNA from any step in a standard ChIP protocol. This includes samples that have undergone reverse cross-linking, Proteinase K or RNase A digestion, mechanical or nuclease-mediated DNA shearing, and samples eluted from chromatin-antibody-bead complexes. The specially formulated ChIP DNA Binding Buffer promotes DNA adsorption to the column in the presence of detergents, antibodies, and proteinases that are often used for ChIP.

  • Two minute DNA clean-up from any step in a standard ChIP protocol.
  • DNA is ideal for PCR, arrays, DNA quantification, Southern blot analysis, sequencing, and other molecular applications.

These kits may be applied to any routine ChIP procedure to determine DNA concentration of samples that have undergone reverse cross-linking following DNA shearing.


Specifications
Elution Volume: ≥ 6 µl
Processing Time: 2 minutes
Recovery: 70-90% for DNA 50 bp - 10 kb, 70% for DNA > 10 kb
Size Range: 50 bp - 23 kb
Storage: All components should be stored at room temperature.

Customers Who Bought This Also Bought