M-Wash Buffer, for use with EZ DNA Methylation Kits, size: 24 ml
Catalog #: 77001-554
Supplier: Zymo Research
M-Wash Buffer, for use with EZ DNA Methylation Kits, size: 24 ml
Catalog #: 77001-554
Supplier: Zymo Research
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$60.20 to $3,424.99
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$260.48 to $788.04
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44