GL45 Cap with NPT Tap, Ace Glass Incorporated
Catalog #: 76107-068
Supplier: Ace Glass
Specifications
- Description:GL45 Cap
- Material:PTFE
- NPT Size:1/4" (Tap)
- Cat. no.:76107-068
Specifications
About this item
GL45 cap with an NPT tap designed to accept various male NPT adapters found, for instance, in ACE compression tube fittings or standard taper joint adapters.
- Caps are made of either polypropylene or PTFE
- Ideal connections for Duran laboratory bottles
- Caps accept various male NPT adapters
Customers Who Bought This Also Bought