Order Entry
ContactUsLinkComponent
Plasmid-Safe* ATP-Dependent Dnase 1000 Units
ATP-Dependent DNase
Catalog #: 76081-626
Supplier:  Lucigen
CAS Number:  
Plasmid-Safe* ATP-Dependent Dnase 1000 Units
ATP-Dependent DNase
Catalog #: 76081-626
Supplier:  Lucigen
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1000 units
  • Enzyme name:
    ATP-Dependent DNase
  • Concentration:
    10 units/μl
  • Cat. no.:
    76081-626

Specifications

About this item

Remove contaminating bacterial chromosomal DNA from plasmid, cosmid, fosmid, and BAC preps without digesting the plasmid, cosmid, fosmid or BAC molecules 

  • Digest contaminating linear DNA without digesting nicked or closed-circular dsDNA or supercoiled DNA (plasmids, fosmids, etc.)
  • Use as a final purification step for plasmid, cosmid, fosmid, and BAC vector and clone preparations
  • Utilize whenever protecting circular dsDNA is important

Plasmid-Safe™ ATP-Dependent DNase selectively removes contaminating bacterial chromosomal DNA from plasmid, cosmid, fosmid, and BAC clones or vector preparations

Plasmid-Safe ATP-Dependent DNase digests linear dsDNA to deoxynucleotides at slightly alkaline pH and, with lower efficiency, closed-circular and linear ssDNA. The enzyme has no activity on nicked or closed-circular dsDNA or supercoiled DNA. Therefore, Plasmid-Safe DNase is ideal as the final purification step for plasmid, cosmid, fosmid, and BAC vector and clone preparations.

Applications include:
Minimizes the possibility of cloning or sequencing contaminating chromosomal DNA from plasmid, cosmid, fosmid, or BAC preparations
Fast and easy protocol with minimal handling time
Complete protocols provided for using Plasmid-Safe DNase with miniprep, midiprep, and maxiprep plasmid, cosmid, fosmid, and BAC DNA purifications

Such preparations are frequently contaminated with fragments of bacterial genomic DNA generated during alkaline lysis. Other purification options, such as spin-columns or even CsCl centrifugation, do not effectively remove these contaminants and require further purification steps. Contaminating DNA fragments left behind by these methods ultimately can become ligated into a cloning vector, resulting in false positives and high backgrounds, or erroneous sequence data.

Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme, 10X reaction buffer and 25 mM ATP.
Customers Who Bought This Also Bought