COD Digestion vials, VWR Chemicals BDH®
Catalog # CABDH0403-150
Supplier: VWR International
CAS Number:
Specifications
- Description:COD Digestion Vials, Hg-free
- Size:150
- Cat. no.:CABDH0403-150
- Range:20 - 1500 mg/L
- Type:Mercury-free, high range
Specifications
About this item
BDH COD Vials used in determination of Chemical Oxygen Demand (COD)
- Easy to Use
- Mercuric formulation follows EPA approved method
- Low Range, High Range, and Mercury Free High Range
Vials contain premeasured digestion reagents suited to your expected range of COD, so there is no measuring of harmful chemicals- simply add your sample to the vial and digest.
Caution: The vials contain hazardous chemicals, typically corrosive, acute toxic and irritant. GHS signal word: Danger
Frequently Bought Together