ContactUsLinkComponent
Cod Digestion Vials, Low Range
COD Digestion vials, VWR Chemicals BDH®
Catalog #: CABDH0400-150
Supplier:  VWR International
CAS Number:  
Cod Digestion Vials, Low Range
COD Digestion vials, VWR Chemicals BDH®
Catalog #: CABDH0400-150
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    COD Digestion Vials
  • Size:
    150
  • Cat. no.:
    CABDH0400-150
  • Range:
    3 - 150 mg/L
  • Type:
    Low range
Specifications
About this item
BDH COD Vials used in determination of Chemical Oxygen Demand (COD)
  • Easy to Use
  • Mercuric formulation follows EPA approved method
  • Low Range, High Range, and Mercury Free High Range
Vials contain premeasured digestion reagents suited to your expected range of COD, so there is no measuring of harmful chemicals- simply add your sample to the vial and digest.
Caution: The vials contain hazardous chemicals, typically corrosive, acute toxic and irritant. GHS signal word: Danger
Customers Who Bought This Also Bought