VWR* Upright Stainless Steel Freezer Drawer Rack 538.5 x 268.5 x 138 mm
Catalog #: 76052-300
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Drawer Rack 538.5 x 268.5 x 138 mm
Catalog #: 76052-300
Supplier: VWR International
Customers Who Bought This Also Bought
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Fused Quartz Microscope Slides and Coverslips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$45.79
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
$99.02 to $485.35