Order Entry
ContactUsLinkComponent
Pulley - Force Table
Force Table Pulley
Catalog #: 470230-108
Supplier:  Wards
Pulley - Force Table
Force Table Pulley
Catalog #: 470230-108
Supplier:  Wards

Specifications

  • Description:
    Force table pulley
  • Cat. no.:
    470230-108

Specifications

About this item

Plastic pulley with table clamp, includes hand knob for tightening.

  • Plastic 2" pulley
  • Clamp made of hard plastic
  • Clamp max opening 1.250"
  • Grey hand knob for tightening clamp

labs low friction force table 2" pulley - Clamp max opening is 1.25" Clamp is made of hard plastic. Grey hand knob for tighten clamp

Ordering information: The corresponding force table (470014-356) is also available for purchase.
Customers Who Bought This Also Bought