ContactUsLinkComponent
Phusion* High-Fidelity PCR Master Mix with HF Buffer 500 reactions
Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Catalog #: CA101641-016
CAS Number:  
Phusion* High-Fidelity PCR Master Mix with HF Buffer 500 reactions
Phusion High-Fidelity PCR Master Mix with HF® Buffer, New England Biolabs
Catalog #: CA101641-016
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Phusion High-Fidelity PCR Master Mix with HF Buffer
  • Size:
    500 Reactions
  • Cat. no.:
    CA101641-016
Specifications
About this item
Phusion High-Fidelity PCR Master Mix with HF Buffer is a 2X master mix consisting of Phusion DNA Polymerase, deoxynucleotides and reaction buffer that has been optimized and includes MgCl2
All that is required is the addition of template, primers and water.
Customers Who Bought This Also Bought