ContactUsLinkComponent
OneTaq* Quick-Load* Master Mix 100 reactions
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Catalog #: CA101640-998
CAS Number:  
OneTaq* Quick-Load* Master Mix 100 reactions
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Catalog #: CA101640-998
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    OneTaq Quick-Load 2X Master Mix with Standard Buffer
  • Size:
    100 Reactions
  • Cat. no.:
    CA101640-998
Specifications
About this item
OneTaq® Quick-Load® 2X Master Mix with Standard Buffer is an optimized, ready-to-use blend of Taq and Deep VentR ™ DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products.
The 3´→5´ exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load master mix formulation contains dNTPs, MgCl2, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.
Customers Who Bought This Also Bought