To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:DNA gel bands
- Sample type:Gel matrix
- Target molecule:DNA
- Description:Zymoclean™ Gel DNA Recovery Kit (capped spin columns)
- Size:50 preps
- Description:Zymoclean™ Gel DNA Recovery Kit
- Yield:50 bp to 23 kb
- Cat. no.:77001-144
Specifications
About this item
The Zymoclean Gel DNA Recovery Kits provide for the rapid purification of high quality DNA from TAE/TBE-bufffered agarose gels.
- Quick (15 minute) recovery of ultra-pure DNA from agarose gels.
- Column design permits DNA elution at high concentrations into minimal volumes (≥ 6 µL).
- Eluted DNA is well suited for use in DNA ligation, sequencing, labeling, PCR, etc.
About Zymoclean™ Gel DNA Recovery Kit
The Zymoclean™ Gel DNA Recovery Kit provides for the rapid purification of high quality DNA from TAE/TBE-buffered agarose gels. The product features Fast-Spin technology to yield high-quality, purified DNA in just minutes. DNA purified using the Zymoclean™ Gel DNA Recovery kit are perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.