DNA Binding Buffer, Size: 100ml
Catalog #: 77001-138
Supplier: Zymo Research
Customers Who Bought This Also Bought
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$135.75 to $468.77
Water, Molecular Biology Grade, sterile RNase Free, DNase Free, Protease Free, DEPC Free
Supplier: Ricca Chemical
$93.96 to $1,045.34
Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$285.47 to $1,142.06