ContactUsLinkComponent
Roll and Grow* Plating Beads 2 tubes
Roll & Grow™ Spherical Glass Plating Beads, MP Biomedicals
Catalog # CA76045-838
Supplier:  MP Biomedicals
undefined
Roll & Grow™ Spherical Glass Plating Beads, MP Biomedicals
Catalog # CA76045-838
Supplier:  MP Biomedicals
Supplier Number:  115000550
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Spherical Glass Plating Beads
  • Cat. no.:
    CA76045-838
  • Packaging:
    Two 50 ml tubes
Specifications
About this item
Roll & Grow™ Plating Beads are used to spread 25–400μL of bacterial or yeast cells across the surface of an agar plate.
  • Sterile
  • Easy to use
  • Spread cells in just seconds per plate
  • Provides even colony distribution
  • Eliminates hazards of flame and ethanol
Roll & Grow™ Plating Beads provide a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow® Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.
Frequently Bought Together