Order Entry
ContactUsLinkComponent
C Ring Blue 1.93in ID
Vinyl-Coated Lead Rings, Argos Technologies
Catalog #: 76040-416
Supplier:  Argos Technologies
C Ring Blue 1.93in ID
Vinyl-Coated Lead Rings, Argos Technologies
Catalog #: 76040-416
Supplier:  Argos Technologies

Specifications

  • Description:
    Blue Vinyl-Coated Lead C-Ring
  • Fits Flask:
    125 - 500 ml
  • O.D.:
    2.99"
  • I.D.:
    1.93"

Specifications

About this item

Perfect for stabilizing glassware and laboratory containers.

  • Chemical-resistant vinyl coating
  • Available in O-shaped or C-shaped rings
  • Optional holder for benchtop organization

Lead Ring Weights are ideal for stabilizing glassware and laboratory containers during water immersion. The vinyl coating prevents scratching on labware or the benchtop. The “C” shaped rings have some flexibility for a more customized fit.

Customers Who Bought This Also Bought