To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Potassium Sodium Tartrate
- Size:500 g
- Chemical/Physical Specifications:Crystal
- Cat. no.:470229-144
- Grade:Laboratory
Specifications
About this item
CAS Number: 6381-59-5
Formula Weight: 282.23
Formula: KNaC4H4O6·4H2O
Hazard Info: Oxidizer, Toxic
Density (g/mL): 2.065
Boiling Point (°C): 105-130
Freezing Point (°C): 36.4
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.