Order Entry
ContactUsLinkComponent
Finnpipette* F2 Variable Volume Single-Channel Pipette 10-100uL
Finnpipette® F2 Single-Channel Variable Volume Pipettors, Thermo Scientific
Catalog #: CA89080-356
Finnpipette* F2 Variable Volume Single-Channel Pipette 10-100uL
Finnpipette® F2 Single-Channel Variable Volume Pipettors, Thermo Scientific
Catalog #: CA89080-356

Specifications

  • Accuracy:
    ±3.0 - 0.8%
  • Volume:
    10 - 100 µl
  • Description:
    Finnpipette® F2 Pipettor
  • Increments:
    0.2 µl
  • Precision:
    1.0–0.2%
  • Color Code:
    Yellow

Specifications

About this item

Finnpipette® F2 variable volume pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures

Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. Each lightweight device offers remarkably smooth pipetting action. Pipettors are fully autoclavable and easy to service. An enlarged digit display improves ease of use, and color coding simplifies instrument selection.

  • Each lightweight device offers remarkably smooth pipetting action
  • Pipettors are fully autoclavable and easy to service

Finnpipette® F2 variable volume pipettors offer outstanding accuracy and precision in laboratory based liquid dispensing procedures

Pipettors are equipped with an advanced volume gearing mechanism (AVG) designed to improve accuracy and precision. An enlarged digit display improves ease of use, and color coding simplifies instrument selection.

These pipettors combine the established Finnpipette® features with state-of-the-art innovations to create a reliable and ergonomic high-performance device.

Ordering information: Finnpipette® F2 devices are supplied with a five-year manufacturer's warranty.
Customers Who Bought This Also Bought