Mix & Go, E. coli Transformation Kit & Buffer Set, Zymo Research
Catalog #: 76020-660
Supplier: Zymo Research
CAS Number:
Specifications
- Description:Mix & Go E. coli Transformation Buffer Kit
- Size:up to 60 ml
- Cat. no.:76020-660
Specifications
About this item
The Mix & Go kit convenient for the preparation of competent E
- It is very simple and efficient DNA transformation
- It completely eliminates the requirement for heat shocking and related procedures
- Transformation efficiencies are typically on the order of 108 – 109 transformants/µg plasmid DNA
The Mix & Go E. coli transformation kit includes all buffers and ZymoBroth™ medium to generate 20 ml of Mix & Go competent cells.
coli cells.
Customers Who Bought This Also Bought