To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:Spin column
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:RNA preps
- Target molecule:RNA
- Description:RNA Clean & Concentrator™-5
- Size:200 Preps
- Description:RNA Clean & Concentrator™
Specifications
About this item
The RNA Clean & Concentrator™ (RCC™) kits facilitate the efficient removal of RNA polymerases, ligases, and RNA modifying enzymes.
- Quick methods for cleaning and concentrating RNA
- Zymo-Spin™ Column/Plate technology allows minimal elution volumes
- Ideal for purification of RNA from aqueous phase following acid phenol extraction
The Zymo RNA Clean and Concentrator™ kits provide simple and reliable methods for the rapid preparation of high-quality RNA. The kit owes its simplicity to a unique single-buffer system and Zymo-Spin™ technology. Simply add the binding buffer to sample, adjust the conditions for binding by adding ethanol, then wash and elute the concentrated RNA. RNA ≥ 17 bases can be safely treated and recovered using these kits. The result is highly concentrated, purified RNA that is perfect for subsequent RNA-based methods, including RT-PCR, hybridization, and more.