Order Entry
ContactUsLinkComponent
RNA Clean & Concentrator*-5 50Preps
RNA Clean & Concentrator™ Kits, Zymo Research
Catalog #: 76020-606
Supplier:  Zymo Research
CAS Number:  
RNA Clean & Concentrator™ Kits, Zymo Research
Catalog #: 76020-606
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Spin column
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    RNA preps
  • Target molecule:
    RNA
  • Description:
    RNA Clean & Concentrator™-5
  • Size:
    50 Preps
  • Description:
    RNA Clean & Concentrator™

Specifications

About this item

The RNA Clean & Concentrator™ (RCC™) kits facilitate the efficient removal of RNA polymerases, ligases, and RNA modifying enzymes.

  • Quick methods for cleaning and concentrating RNA
  • Zymo-Spin™ Column/Plate technology allows minimal elution volumes
  • Ideal for purification of RNA from aqueous phase following acid phenol extraction

The Zymo RNA Clean and Concentrator™ kits provide simple and reliable methods for the rapid preparation of high-quality RNA. The kit owes its simplicity to a unique single-buffer system and Zymo-Spin™ technology. Simply add the binding buffer to sample, adjust the conditions for binding by adding ethanol, then wash and elute the concentrated RNA. RNA ≥ 17 bases can be safely treated and recovered using these kits. The result is highly concentrated, purified RNA that is perfect for subsequent RNA-based methods, including RT-PCR, hybridization, and more.

Customers Who Bought This Also Bought