DNA Wash Buffer, Concentrate, Zymo Research
Catalog #: 76020-534
Supplier: Zymo Research
CAS Number:
Specifications
- Description:DNA Wash Buffer
- Size:24 ml
- Cat. no.:76020-534
Specifications
About this item
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
NEB®5-alpha Competent <i>E.coli</i> (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
$300.00