Order Entry
ContactUsLinkComponent
Direct-zol* RNA MicroPrep 50 Preps
Direct-zol™ RNA Kits, Zymo Research
Catalog #: 76020-456
Supplier:  Zymo Research
CAS Number:  
Direct-zol™ RNA Kits, Zymo Research
Catalog #: 76020-456
Supplier:  Zymo Research
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Direct-zol™ RNA Microprep Kit
  • Size:
    50 Preps
  • Sample size:
    ≤1×10Cells
  • Description:
    Direct-zol™ RNA Microprep Kit
  • Sample capacity:
    10 µg
  • Cat. no.:
    76020-456

Specifications

About this item

RNA from TRIzol® in seven minutes.

  • Extract RNA from any sample (cells, tissues, blood, biological liquids, etc.)
  • Easy Handling: No phase separation or precipitation steps
  • NGS-ready: Ultra-pure RNA without phenol carryover, no DNA contamination (DNase I included)
  • Total RNA Recovery: All RNA >17 nt is recovered

The Direct-zol™ RNA Kits provide a streamlined method for the purification of up to 100 µg (per prep) of high-quality RNA directly from samples in TRI Reagent® or similar. Total RNA, including small RNAs (17 to 200 nt), is effectively isolated from a variety of sample sources (cells, tissues, serum, plasma, blood, biological liquids, etc.).

The procedure is easy. Simply apply a prepared sample in TRI Reagent® directly to the Zymo-Spin™ column and then bind, wash, and elute the RNA. No phase separation, precipitation, or post-purification steps are necessary. The eluted RNA is high quality and suitable for subsequent molecular manipulation and analysis (including RT-PCR, transcription profiling, hybridization, sequencing and more).

Customers Who Bought This Also Bought