DNA/RNA Shield™ Reagent, Zymo Research
Catalog #: 76020-440
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Chemical extraction & preservation
- Prep size:Scalable
- Refine target molecule:Total RNATotal DNA
- Sample type:StoolSwabsCultured cellsHuman/animal cells and tissues
- Target molecule:Copurification of DNA & RNADNARNA
- Description:DNA/RNA Shield™ (2X Concentrate)
- Size:125 ml
- Description:DNA/RNA Shield™ (2X Concentrate)
- Cat. no.:76020-440
Specifications
About this item
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.
- Nucleic acid preservation (at ambient temperature; cold-free)
- Pathogen inactivation (bacteria, fungus, parasites & viruses)
- RNA Stability: Ambient temperature (4 to 25 °C) >1 month
- DNA Stability: Ambient temperature (4 to 25 °C) >2 years
- DNA and RNA: Frozen (<−20 °C): Indefinitely
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialized equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).
Customers Who Bought This Also Bought