To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:Quick-RNA™ Midiprep Kit
- Size:25 Preps
- Sample size:≤1×108 Cells or 100 mg Tissue
- Description:Quick-RNA™ Midiprep Kit
- Sample capacity:1000 µg
- Yield:Midiprep: 1 mg RNA (binding capacity), ≥ 200 µl (elution volume)
- Cat. no.:76020-130
Specifications
About this item
Isolation of RNA from cells and soft tissues.
- DNA-free: Genomic DNA removal column and DNase I included
- NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more
The quick-RNA kits are fast RNA isolation kits designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell and tissue samples. The procedure combines a unique buffer system with zymo-spin column technology to yield high quality total RNA (including small RNAs 17 to 200 nt) in about 10 minutes.
The procedure for these RNA isolation kits is simple and fast. Add the provided RNA lysis buffer to a sample, and then purify the RNA using the zymo-spin columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more.
In addition, these RNA isolation kits can be used for the enrichment of small and large RNAs into separate fractions.