Preserved Turtles
Catalog #: 470228-314
Supplier: Avantor
Specifications
- Description:Formalin Preserved Turtles
- Size:8 to 10"
- Injection Type:Double injected
- Cat. no.:470228-314
- Packaging:Pail of 10
Specifications
About this item
Formalin preserved turtles available in pails.
- Reptile anatomy
- General dissection
- High quality preservation
We offer Pseudemys sp. turtles for study specimens that can be used to teach comparative anatomy and the similarities and differences between reptiles, amphibians, and mammals. Students can explore the structures of the shell, musculature, and internal organs of the study animal. The carapace is pre-cut for ease of dissection. We offer plain preserved, single, double, or triple injected turtles for your teaching or research needs.
Customers Who Bought This Also Bought