Polystyrene Reagent Reservoirs
Catalog #: 470228-462
Supplier: MTC BIO, INC.
Specifications
- Capacity:50 ml
- Length:152 mm
- Sterility:Sterile
- Width:86 mm
- Description:Reagent Reservoir
- Height:29 mm
- Cat. no.:470228-462
Specifications
About this item
Standard 4-channel, 8-channel, 12-channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pouring, all versions have independent spouts in each corner. Bottoms are formed with a sharply tapered 'V' shape to minimize waste and reagent consumption.
- Graduated
- Anti-static
- Sharp "V" Bottoms
- Pouring spout on all 4 corners
- Divided options are available
- All reservoirs are RNAse- and DNAse-free
Customers Who Bought This Also Bought