CoolCaddy™ Cold WorkStation
Catalog #: 470228-446
Supplier: MTC BIO, INC.
Specifications
- Description:CoolCaddy™ cold workstation
- Cat. no.:470228-446
- Dimensions:21×17×4.5 cm
Specifications
About this item
Keeps your samples cool while you experiment.
- Maintains 1 to 4 °C for up to 5 hours
- Keeps samples cool but not frozen
- Eliminates the need for crushed ice
- Accomodates most small tube types
Keeps your reagents and samples cool on the lab bench without the use of crushed ice.
Simply freeze overnight and its ready to hold 1 to 4 °C for up to 5 hours, keeping your Microtubes, Cryovials, PCR Tubes/Plate and Reagent Reservoirs cold (but not frozen) while you work.
Customers Who Bought This Also Bought