Order Entry
ContactUsLinkComponent
Trypsin-ultra* Mass Spectrometry Grade 100 ug
Bovine Trypsin (from Pancreas)
Catalog #: CA101228-584
CAS Number:  
Trypsin-ultra* Mass Spectrometry Grade 100 ug
Bovine Trypsin (from Pancreas)
Catalog #: CA101228-584
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    Pancreas
  • Species:
    Bovine
  • Size:
    100 µg
  • Storage conditions:
    -20 °C
  • Enzyme name:
    Trypsin (TPCK-treated)
  • Enzyme activity:
    2.1 µmol/min/mg
  • Molecular weight:
    Theoretical: 23675 Da
  • Grade:
    Mass Spectrometry Grade
  • Tested applications:
    Protein Digestion, Proteases
  • Cat. no.:
    CA101228-584

Specifications

About this item

Trypsin-ultra™, Mass Spectrometry Grade is a serine endopeptidase

  • Digestion of proteins for proteomic analysis by Mass Spectrometry
  • Protein and peptide identification
  • Concentration 100 ng/µL

It selectively cleaves peptide bonds C-terminal to lysine and arginine residues. Trypsin-ultra is treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inactivate any remaining chymotryptic activity. It is modified by acetylation of the ε-amino groups of lysine residues to prevent autolysis. Trypsin-ultra (TPCKtreated) cleaves at Lys-Pro and Arg-Pro bonds at a much slower rate than other amino acid residues. Isolated from bovine (Bos taurus) pancreas.

Packaging: The Trypsin-ultra, Reaction Buffer is included with this product.
Customers Who Bought This Also Bought