ContactUsLinkComponent
DNA Ladder 100 bp
DNA Ladder, 100 bp, New England Biolabs
Catalog #: CA101228-490
CAS Number:  
DNA Ladder 100 bp
DNA Ladder, 100 bp, New England Biolabs
Catalog #: CA101228-490
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    100 bp DNA Ladder
  • Size:
    100 gel lanes
  • Cat. no.:
    CA101228-490
Specifications
About this item
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis.
The digested DNA includes fragments ranging from 100 to 1517 base pairs. The 500 and 1000 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each of the bands is provided (assuming a 0.5 μg load) for approximating the mass of DNA in comparably intense samples of similar size.
Delivery: Supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Customers Who Bought This Also Bought