ContactUsLinkComponent
Monarch* Plasmid Miniprep Kit 50 preps
Monarch™ Plasmid Miniprep Kit and Components, New England Biolabs®
Catalog #: CA102971-698
   Sustainable Packaging
   Sustainable Materials
CAS Number:  
Monarch* Plasmid Miniprep Kit 50 preps
Monarch™ Plasmid Miniprep Kit and Components, New England Biolabs®
Catalog #: CA102971-698
   Sustainable Packaging
   Sustainable Materials
CAS Number:  
Specifications
  • Description:
    Monarch Plasmid Miniprep Kit
  • Environmentally Preferable:
    Sustainable Packaging
    Sustainable Materials
  • Size:
    50 Preps
  • Cat. no.:
    CA102971-698
Specifications
About this item
The Monarch Plasmid Miniprep Kit is a rapid and reliable method for the purification of high quality plasmid DNA.
  • Elute in as little as 30 μl
  • Eliminate buffer retention and salt carryover with optimized column design
  • Save time with fast, user-friendly protocols
  • Easily label columns using tab and frosted surfaces
  • Monitor completion of certain steps and easily identify buffers with colored buffer system
  • No need to add RNase before starting
  • Reduced environmental impact
  • Recyclable packaging
This method employs standard cell resuspension, alkaline lysis, and neutralization steps, with the additional benefit of color indicators at certain steps to easily monitor completion. Unique wash buffers ensure salts, proteins, RNA and other cellular components are removed, allowing low-volume elution of concentrated, highly pure DNA. Protocols are fast and user friendly, saving you valuable time. Elution in as little as 30 μl provides concentrated DNA for use in downstream applications, such as restriction digests, DNA sequencing, PCR and other enzymatic manipulations. Designed with sustainability in mind, Monarch kits use significantly less plastic and responsibly-sourced, recyclable packaging.
Customers Who Bought This Also Bought