Order Entry
ContactUsLinkComponent
NEBNext* FFPE DNA Repair Mix 24 reactions
NEBNext® NGS DNA Library Preparation for Illumina, Kits and Products, New England Biolabs
Catalog #: CA102902-482
CAS Number:  
NEBNext* FFPE DNA Repair Mix 24 reactions
NEBNext® NGS DNA Library Preparation for Illumina, Kits and Products, New England Biolabs
Catalog #: CA102902-482
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    NEBNext® FFPE DNA repair mix
  • No. of Reactions:
    24

Specifications

About this item

The NEBNext Ultra II Q5® Master Mix is a new formulation of Q5 DNA polymerase that has been optimized for robust, high-fidelity amplification of next-generation sequencing (NGS) libraries.

  • Fast, streamlined workflow and novel master mixes
  • High yield preparation of high quality libraries
  • Functionally validated by preparation of a library

This new formulation further improves the uniformity of amplification of libraries, including superior performance with GC-rich regions

12-fold higher than that of Pfu DNA Polymerase), resulting in ultra-low error rates. The NEBNext Ultra II Q5 Master Mix is an aptamer-based hot start formulation that allows convenient room temperature reaction set up. The convenient 2X master mix format contains dNTPs, Mg++ and a proprietary buffer. NEBNext Ultra II Q5 Master Mix is also included in the NEBNext Ultra II DNA Library Prep Kit for Illumina®.

Get more of what you need, with the highest library yields. Use these products to generate high quality libraries even when you have only limited amounts of DNA, with inputs as low as 500 pg (Ultra II). Prepare libraries from ALL of your samples, including GC-rich targets and FFPE DNA samples. Improve yields and quality for target enrichment applications. Save time with streamlined workflows, reduced hands-on time, and automation compatibility.

Q5 High-Fidelity DNA Polymerase is fused to a processivity-enhancing Sso7d domain providing the highest fidelity available (

Certifications: In addition to the extensive QCs on individual kit components, all NEBNext kits for Illumina are functionally validated by preparation of a library, followed by Illumina sequencing.
Ordering information: Original NEBNext Ultra kits and sets of individual reagents are also available.
Caution: Fast, streamlined workflow and novel master mixes High yield preparation of high quality libraries Functionally validated by preparation of a library
Customers Who Bought This Also Bought