Order Entry
ContactUsLinkComponent
Gel Loading Dye, Purple (6X) 4 ml
Gel Loading Dye Purple (6X), New England Biolabs
Catalog #: CA102877-610
CAS Number:  
Gel Loading Dye, Purple (6X) 4 ml
Gel Loading Dye Purple (6X), New England Biolabs
Catalog #: CA102877-610
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Gel Loading Dye Purple (6X)
  • Size:
    4.0 mL

Specifications

About this item

Gel Loading Dye, Purple (6X) is a pre-mixed loading buffer which contains a combination of two dyes, Dye 1 (pink/red) and Dye 2 (blue).

The red dye serves as the tracking dye for both agarose and non-denaturing polyacrylamide gel electrophoresis. The two dyes separate upon gel electrophoresis; the red band is the major indicator and migrates similarly to Bromophenol Blue on agarose gels. Specifically chosen, this dye does not leave a shadow under UV light. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. The dye also contains Ficoll®, which creates brighter and tighter bands when compared to glycerol loading dyes.

Customers Who Bought This Also Bought