Order Entry
ContactUsLinkComponent
NEB* PCR Cloning Kit 20 reactions
NEB® PCR Cloning Kit, New England Biolabs
Catalog #: CA102877-564
CAS Number:  
NEB* PCR Cloning Kit 20 reactions
NEB® PCR Cloning Kit, New England Biolabs
Catalog #: CA102877-564
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    NEB PCR Cloning Kit
  • Cat. no.:
    CA102877-564
  • No. of Reactions:
    20

Specifications

About this item

This PCR Cloning Kit contains an optimized 2X Cloning Master Mix with a proprietary ligation enhancer and a linearized vector that uses a novel mechanism for background colony suppression to give a low background.

It allows simple and quick cloning of any PCR amplicon, whether the amplification reactions are performed with proofreading DNA polymerases, such as Q5® or Phusion® which produce blunt ends; or nonproofreading DNA polymerases, such as Taq or Taq mixes (OneTaq®, LongAmp™ Taq) which produce single base overhangs. This is possible due to “invisible” end polishing components in the master mix that are active during the ligation step. The kit also allows direct cloning from amplification reactions without purification, and works well whether or not the primers used in the PCR possess 5´-phosphate groups.

Customers Who Bought This Also Bought