ContactUsLinkComponent
Remove-iT* PNGase F 6,750 units
Remove-iT PNGase F
Catalog #: CA102855-176
CAS Number:  
Remove-iT* PNGase F 6,750 units
Remove-iT PNGase F
Catalog #: CA102855-176
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    6750 units
  • Enzyme name:
    Remove-iT PNGase F
  • Cat. no.:
    CA102855-176
Specifications
About this item
Remove-iT® PNGase F is an amidase which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins
Remove-iT PNGase F is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.
Customers Who Bought This Also Bought