Order Entry
ContactUsLinkComponent
Q5* High-Fidelity PCR Kit 50 reactions
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Catalog #: CA102855-120
CAS Number:  
Q5* High-Fidelity PCR Kit 50 reactions
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Catalog #: CA102855-120
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
  • Description:
    Q5® High-Fidelity PCR kit
  • Size:
    50 Reactions
  • Cat. no.:
    CA102855-120

Specifications

About this item

The Q5® High-Fidelity 2X Master Mix offers robust, high-fidelity performance in a convenient master mix format.

  • For use in high-specificity PCR, high-fidelity PCR, cloning, long or difficult amplification and high-throughput PCR.

The Q5 High- Fidelity 2X Master Mix features a high-fidelity, thermostable DNA polymerase with 3´→ 5´ exonuclease activity, fused to a processivity-enhancing Sso7d domain to support robust DNA amplification. With an error rate > 100-fold lower than that of Taq DNA Polymerase and 12-fold lower than that of Pyrococcus furiosus (Pfu) DNA Polymerase, Q5 High-Fidelity DNA Polymerase is ideal for cloning and can be used for long or difficult amplicons. The convenient master mix formulation is supplied at a 2X concentration. The mix contains dNTPs, Mg++ and a proprietary broad-use buffer requiring only the addition of primers and DNA template for robust amplification regardless of GC content. When used at the recommended 1X final concentration, the Q5 High-Fidelity Master Mix contains 2 mM MgCl2. Q5 High-Fidelity DNA Polymerase is unlike typical, lower fidelity PCR enzymes.

Ordering information: The Q5 High-Fidelity DNA Polymerase and the Q5 Hot Start High-Fidelity DNA Polymerase are supplied with Q5 High GC Enhancer and Q5 Reaction Buffer.
Customers Who Bought This Also Bought