ContactUsLinkComponent
Blunt/TA Ligase Master Mix 50reactions
Blunt/TA Ligase Master Mix, New England Biolabs
Catalog #: CA102715-940
CAS Number:  
Blunt/TA Ligase Master Mix 50reactions
Blunt/TA Ligase Master Mix, New England Biolabs
Catalog #: CA102715-940
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Blunt/TA Ligase Master Mix
  • Size:
    50 Reactions
  • Cat. no.:
    CA102715-940
Specifications
About this item
Blunt/TA Ligase Master Mix is a ready-to-use solution of T4 DNA Ligase, proprietary ligation enhancer, and optimized reaction buffer.
This master mix is specifically formulated to improve ligation and transformation of both blunt-end and single-base overhang substrates. The master mix format simplifies reaction set-up, ensures an optimized ratio of enzyme and buffer components, and yields robust, rapid ligation of all types of DNA ends using a short incubation time at room temperature. No thawing is necessary as it remains liquid during storage at -20°C. Ligations for subcloning can be carried out in small volumes with low DNA concentrations, allowing users to conserve precious DNA samples and directly transform many strains of chemically competent E. coli without dilution.
Customers Who Bought This Also Bought