Order Entry
ContactUsLinkComponent
NotI-HF* (High Fidelity Enzyme) 500 units
NotI-HF®, Restriction Enzymes, New England Biolabs
Catalog #: CA101447-642
CAS Number:  
NotI-HF* (High Fidelity Enzyme) 500 units
NotI-HF®, Restriction Enzymes, New England Biolabs
Catalog #: CA101447-642
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    20000 U/ml
  • Description:
    NotI-HF®
  • Size:
    500 units
  • Enzyme name:
    NotI-HF
  • Cat. no.:
    CA101447-642

Specifications

About this item

An E.coli strain that carries the cloned and modified NotI gene from Nocardia otitidis-caviarum (ATCC 14630).

High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. NotI HF® is engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.

Customers Who Bought This Also Bought