AMV Reverse Transcriptase, New England Biolabs
Catalog #: CA101417-976
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:AMV Reverse Transcriptase
- Size:200 units
- Cat. no.:CA101417-976
Specifications
About this item
Avian Myeloblastosis Virus (AMV) Reverse Transcriptase is an RNA-directed DNA polymerase
This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template.
Customers Who Bought This Also Bought