NEB®10-beta Competent E.coli (High Efficiency), New England Biolabs
Catalog #: CA101417-920
Supplier: New England Biolabs (NEB)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:NEB 10-beta competent E.coli (high efficiency)
- Size:6×0.2 ml
- Cat. no.:CA101417-920
Specifications
About this item
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high-quality plasmid preparations.
- High efficiency strain ideal for cloning large plasmids and BACs
- Available in single-use vials, 200 µl vials, and electrocompetent format
- No dry ice surcharge on competent cell shipments
- Outgrowth medium included
- Free of animal products
Customers Who Bought This Also Bought