ContactUsLinkComponent
DNA Ladder 0.1 ml
DNA Ladder, 50 bp, New England Biolabs
Catalog #: CA101228-506
CAS Number:  
DNA Ladder 0.1 ml
DNA Ladder, 50 bp, New England Biolabs
Catalog #: CA101228-506
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    50 bp DNA Ladder
  • Size:
    200 gel lanes
  • Cat. no.:
    CA101228-506
Specifications
About this item
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 17 bands suitable for use as molecular weight standards for agarose and polyacrylamide gel electrophoresis
The digested DNA includes fragments ranging from 50-1,350 base pairs. The 200 and 500 base pair bands have increased intensity to serve as reference points. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Customers Who Bought This Also Bought