ContactUsLinkComponent
ssRNA Ladder 25 gel lanes
ssRNA Ladder, New England Biolabs
Catalog #: CA101228-410
CAS Number:  
ssRNA Ladder 25 gel lanes
ssRNA Ladder, New England Biolabs
Catalog #: CA101228-410
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ssRNA Ladder
  • Size:
    25 gel lanes
  • Cat. no.:
    CA101228-410
Specifications
About this item
The ssRNA Ladder is a set of 7 RNA molecules produced by in vitro transcription of a mixture of 7 linear DNA templates
The ladder sizes are: 9000, 7000, 5000, 3000, 2000, 1000 and 500 bases. The 3000 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing or native agarose gels.
Customers Who Bought This Also Bought